Cole-Parmer Polycarbonate Specific Gravity Hydrometers, plain form
These durable polycarbonate hydrometers won't break when dropped—perfect for industrial or laboratory use.
The temperature standard for these hydrometers is 60°F (15°C). All hydrometers are individually serialized, feature metal ballast with wax epoxy and include multi-language statement of accuracy supplied by the vendor.
Note: Polycarbonate hydrometers are not recommended for use with petroleum-based liquids or other corrosives that would potentially affect the plastic material. They should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.
ITEM(S) ON SALE
$103.91 - $118.78USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
1.000/1.180 Specific Gravity Shatterproof Plastic Hydrometer
$103.91USD / Each
$122.25USD / Each
COMPARE
0.920/1.080 Specific Gravity Shatterproof Plastic Hydrometer
$112.62USD / Each
$132.50USD / Each
COMPARE
1.050/1.220 Specific Gravity Shatterproof Plastic Hydrometer
$112.41USD / Each
$132.25USD / Each
COMPARE
1.200/1.420 Specific Gravity Shatterproof Plastic Hydrometer
$112.41USD / Each
$132.25USD / Each
COMPARE
1.400/1.620 Specific Gravity Shatterproof Plastic Hydrometer
$118.78USD / Each
$139.75USD / Each