This product is now discontinued, View Related Products
Cole-Parmer® PBH High-Capacity Portable Toploading Balances with NTEP Approval
Easily weigh heavy dynamic loads
  • Included software collects and exports data for further analysis
  • Models with NTEP approval are available for commercial applications—legal for trade
  • Built-in rechargeable battery provides portability
  • Connect to printer or PC with RS-232 interface
  • Weighing modes include parts counting, percent weighing, totalizing, check weighing, peak hold, and animal weighing
  • 5-year industry-leading warranty
These toploading balances feature high weighing capacities and the ability to weigh moving samples, such as live animals. Included data collection software allows you to export logged data for further analysis, and RS-232 interface provides printer or PC connection. Balances may run on mains power, or use the built-in rechargeable battery where portability is necessary.
Models with NTEP approval should be selected if your weighing application includes buying, selling, or charging based upon weight within the United States. These models have been tested and proven to meet all government standards and requirements and are considered legal for trade. . Each model features a “d” and “e” value. The “d” value is defined in NIST Handbook 44 as the scale division, value of (d) and is the smallest division a balance can display. The “e” value is known as the verification scale division, value of (e). It represents the stated accuracy and is utilized as the value for commercial transactions. Refer to your state guidelines on “d” vs “e” value acceptance.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Calibration Type Capacity (g) Description
Availability Pricing
COMPARE
External 6000 High Capacity Portable Toploading Balance, d=0.1g, e=1g (see text for details)
Call For Price
COMPARE
Internal 1000 High Capacity Portable Toploading Balance, d=0.01g, e=1g (see text for details)
Call For Price
COMPARE
Internal 2000 High Capacity Portable Toploading Balance, d=0.01g, e=1g (see text for details)
Call For Price
COMPARE
Internal 6000 High Capacity Portable Toploading Balance, d=0.1g, e=1g (see text for details)