Cole-Parmer Microplate Shakers
Mix micro-scale volumes effectively
  • Digital display ensures reproducible results
  • Platform with lip and nonskid mat holds plates securely
  • Set built-in timer for up to 9999 minutes or use continuous mode for added versatility
Designed for microwell plates, this shaker operates with a small orbit at high speed. Program a saved time and speed profile for routine tasks with the user-friendly interface.
Choose the four-plate model (51900-55 and -57) for personal use or for incubators and fume hoods. The eight-plate model (51900-51 and -53) is ideal for large labs and high-throughput applications. Optional tube adapters hold microcentrifuge tubes (order separately).
$825.75 - $1,621.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Load (lbs) Power (VAC)
Availability Pricing
COMPARE
4.4 120
$1,621.00USD / Each
COMPARE
4.4 230
$1,530.00USD / Each
COMPARE
2.2 230
$825.75USD / Each
COMPARE
2.2 120
$855.00USD / Each