This product is now discontinued, View Related Products
Cole-Parmer Magnetic Stirring Hot Plate
Stepless speed control provides smooth stirring action to 1500 rpm
  • Unique U-shaped magnet holds stir bar tightly
Use our economical stirrer for all of your magnetic stirring applications—from gentle agitating to vigorous churning. The large, permanent drive magnet ensures positive coupling of drive to stir bar. Permanently lubricated, brushless drive motor provides long, trouble-free operation. Stirring hot plates are available wtih anodized aluminum or ceramic top plates. Choose the corrosion-resistant ceramic for your higher temperature applications.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Top Plate Length (cm) Top Plate Material
Availability Pricing
COMPARE
115 15.2 Aluminum
Call For Price
COMPARE
230 15.2 Aluminum
Call For Price
COMPARE
115 15.2 Ceramic
Call For Price
COMPARE
230 15.2 Ceramic
Call For Price
COMPARE
115 30.5 Aluminum
Call For Price
COMPARE
230 30.5 Aluminum
Call For Price
COMPARE
115 30.5 Ceramic
Call For Price
COMPARE
230 30.5 Ceramic
Call For Price
Showing 1- 8 of 8