Cole-Parmer® LB-400 Series Precision Toploading NTEP-Approved Balances
Quality and precision at an economical price
  • Designed and engineered to meet stringent NTEP requirements for use in legal applications such as pharmacies, jewelry stores and retail outlets
  • 5-year industry-leading warranty
  • Large, backlit LCD provides clear visibility of weighing results
  • Automatic and manual internal calibration options
  • Removable draft shield on milligram (0.001g) readability options
  • USB, RS-232, Ethernet, and Wi-Fi interfaces
  • Filter settings can be easily adjusted depending on the working environment
  • Multiple working modes handle a variety of weighing functions
The Cole-Parmer LT series precision toploading balances provide advanced weighing features at an economical price. The large, stainless steel weigh pan allows for easy weighing of large items. Easy to operate, the balances can be automatically or manually calibrated internally. The large, backlit LCD allows a clear presentation of the weighing results.
To achieve the best possible weighing conditions, the balances feature a filter level setting that can be adjusted to a fast or slow mode depending on the working environment. Other features include quick access keys, balance taring, balance zeroing, auto switch-off, and backlight turn-off time.
The balance features several databases for recording users, products, weighings, and tares. Only users with appropriate access can adjust balance settings, export data, import data, and delete records. Two USB, two RS-232 interfaces, Ethernet, and
These NTEP-approved balances are tested and evaluated to ensure they meet all government standards and requirements set by the National Institute of Standards and Technology (NIST). Each model features a “d” and “e” value. The “d” value is defined in NIST Handbook 44 as the scale division, value of (d) and is the smallest division a balance can display. The “e” value is known as the verification scale division, value of (e). It represents the stated accuracy and is utilized as the value for commercial transactions. Refer to your state guidelines on “d” vs “e” value acceptance.
ITEM(S) ON SALE
$1,653.52 - $2,068.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g) Description
Availability Pricing
COMPARE
360 0.001 Precision Toploading Balance with LCD, d=0.001g, e=0.01g (see text for details)
$1,732.72USD / Each
$1,969.00USD / Each
COMPARE
1000 0.001 Precision Toploading Balance with LCD, d=0.001g, e=0.01g (see text for details)
$2,068.00USD / Each
$2,350.00USD / Each
COMPARE
2100 0.01 Precision Toploading Balance with LCD, d=0.01g, e=0.1g (see text for details)
$1,653.52USD / Each
$1,879.00USD / Each
COMPARE
4500 0.01 Precision Toploading Balance with LCD, d=0.01g, e=0.1g (see text for details)
$1,804.88USD / Each
$2,051.00USD / Each