Cole-Parmer® Essentials Humidity Sponges and Desiccant Bags
Provides up to 37% moisture absorption
  • Regenerable bags can be regenerated and reused repeatedly
  • Indicators show when moisture capacity has been reached
Humidity sponges meet all drying requirements. One bag will retain 16% moisture in a container with a volume of two gallons (476 cu. in.) The sponges are non-toxic and are Class M3.5 cleanroom-compatible.
$56.00USD / Pkg of 40
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Exterior Depth (in) Exterior Width (in) Description
Availability Pricing
COMPARE
3 3 Humidity Sponge, Indicating; 3\" X 3\" bags, 40 bags/pk
$56.00USD / Pkg of 40
COMPARE
3 3 Humidity Sponge, Regenerable; 3\" X 3\" bags, 40 bags/pk
$56.00USD / Pkg of 40