Cole-Parmer­­® High-Purity Micro-Quartz Filter Sheets
The right choice for testing hot stack gases and filtration demanding high purity media
  • Ideal for low particle levels
  • Made of extremely pure micro-quartz fibers
Laboratories and other stringent environments can count on the purity of these binder-free filter sheets, which have been conditioned by high temperature preheating. Grade MK 5 sheet is suitable for air monitoring applications in high temperatures, aggressive atmospheres and when low levels of trace elements are essential. Grade MK 360 sheet featuring 100% quartz microfibers is for applications with even higher temperatures (500 to 900°C) demanding extremely low levels of trace elements.
$352.50 - $466.25USD / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Sheet Length (in) Sheet Width (in) Description
Availability Pricing
COMPARE
10 8 Grade MK5 High-Purity Quartz Filter Sheet, 8\" x 10\", 932°F (500°C); 25/Pk
$352.50USD / Pkg of 25
COMPARE
10 8 Grade MK360 High-Purity Quartz Filter Sheet, 8\" x 10\", 1652°F (900°C); 25/Pk
$466.25USD / Pkg of 25