Cole-Parmer High-Accuracy Gauge Pressure Transmitters
Welded stainless steel (SS) housing rated to NEMA 4 (IP65) standards
Transducers feature SS capacitive sensor for your more demanding systems—even ElectroMagnetic Interference (EMI)/Radio Frequency Interference (RFI) resistance ensures accurate and consistent performance in any environment.
Transducers feature adjustable zero and span potentiometers. Transducers with 4 to 20 mA output have red (positive power) and black (output) wires. Transducers with 0.1 to 5.1 V output have red (positive power), green (output), and white/black (common) wires.
ITEM(S) ON SALE
$430.10USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Output Resolution
Availability Pricing
COMPARE
25 4 to 20 mA
$430.10USD / Each
$506.00USD / Each
COMPARE
50 4 to 20 mA
$430.10USD / Each
$506.00USD / Each
COMPARE
100 4 to 20 mA
$430.10USD / Each
$506.00USD / Each
COMPARE
500 4 to 20 mA
$430.10USD / Each
$506.00USD / Each
COMPARE
3000 4 to 20 mA
$430.10USD / Each
$506.00USD / Each
COMPARE
25 0.1 to 5.1 VDC
$430.10USD / Each
$506.00USD / Each
COMPARE
500 0.1 to 5.1 VDC
$430.10USD / Each
$506.00USD / Each
COMPARE
3000 0.1 to 5.1 VDC
$430.10USD / Each
$506.00USD / Each
Showing 1- 8 of 8