Cole-Parmer GP Laboratory Overhead Mixer
Mixer designed for reliability and durability
  • Ideal for light-duty mixing of water-like fluids
  • Brushed DC motor with integrated overload protection
  • Durable—no wear parts that require user service
  • IP44 rated per IEC 60529
  • User-replaceable keyless chuck
Cole-Parmer GP laboratory mixer ensures accurate, dependable motor speed control with “soft-start” feature and smooth, gradual adjustment to the speed setting. Constant speed is maintained electronically via automatic compensation for changes in load torque demands at both high and low speeds. Built-in motor current compensation minimizes speed drift with changes in load. Motor current limiting protects the system in case of a locked rotor condition. Output circuit protection protects against inadvertent output shorting.
The mixer rotates at 40 to 4000 rpm with a maximum torque of 9.5 in-oz (6.7 N·cm). The speed control knob is used to turn on and set the speed of the mixer motor and provides a 100:1 adjustment range. The LED indicates the mixer is powered on when it is green. When it flashes, it indicates a limit condition has been reached, such as an overtemperature or over-current situation.
ITEM(S) ON SALE
$1,288.00 - $1,310.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Min Speed (rpm) Description
Availability Pricing
COMPARE
4000 40 GP Mixer Set; 120/230 VAC
$1,310.00USD / Each
COMPARE
4000 40 GP Mixer System; 120/230 VAC
$1,288.00USD / Each
$1,935.00USD / Each