Cole-Parmer Gauge Pressure Transducers, 316 SS
Perfect for high-purity gas delivery systems, semiconductor processes, and control applications that require ultraclean operation and exceptional long-term stability
  • Ideal for demanding applications that require corrosion resistance
  • Features ElectroMagnetic Interference (EMI) and Radio Frequency Interference (RFI) shielding where noise could interrupt a transmitter signal
Rotate cover to access zero and span potentiometers. Transmitters with 4 to 20 mA output have red (positive power) and black (output) wires. Transmitters with 0.2 to 5.2 V output have red (positive power), green (output), and black (common) wires.
ITEM(S) ON SALE
$719.10USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Output
Availability Pricing
COMPARE
250 4 to 20 mA
$719.10USD / Each
$846.00USD / Each
COMPARE
500 4 to 20 mA
$719.10USD / Each
$846.00USD / Each
COMPARE
25 0.2 to 5.2 V
$719.10USD / Each
$846.00USD / Each
COMPARE
100 0.2 to 5.2 V
$719.10USD / Each
$846.00USD / Each