Cole-Parmer® elements™ Glass Erlenmeyer Flasks
Economical option for everyday use flasks
  • Easy read graduations
  • Withstands wide temperature range -50 to 550ºC
  • Graduated tolerances to ±5%
  • Autoclavable
Made with durable borosilicate 3.3 glass and featuring a heavy-duty rim with narrow-mouth opening there are very few applications these flasks cannot handle.
$10.80 - $34.25USD / Pkg of 12
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Description
Availability Pricing
COMPARE
25 5 Erlenmeyer Flask, Glass, 25 mL, 12/pk
$19.50USD / Pkg of 24
COMPARE
50 10 Erlenmeyer Flask, Glass, 50 mL, 12/pk
$16.50USD / Pkg of 12
COMPARE
100 20 Erlenmeyer Flask, Glass, 100 mL, 12/pk
$18.30USD / Pkg of 12
COMPARE
125 20 Erlenmeyer Flask, Glass, 125 mL, 12/pk
$18.80USD / Pkg of 12
COMPARE
250 50 Erlenmeyer Flask, Glass, 250 mL, 12/pk
$25.50USD / Pkg of 12
COMPARE
300 50 Erlenmeyer Flask, Glass, 300 mL, 12/pk
$16.80USD / Pkg of 8
COMPARE
500 100 Erlenmeyer Flask, Glass, 500 mL, 8/pk
$25.50USD / Pkg of 8
COMPARE
1000 100 Erlenmeyer Flask, Glass, 1000 mL, 6/pk
$34.25USD / Pkg of 6
COMPARE
2000 400 Erlenmeyer Flask, Glass, 2000 mL, 1/pk
$10.80USD / Pkg of 1
COMPARE
3000 500 Erlenmeyer Flask, Glass, 3000 mL, 1/pk
$16.30USD / Pkg of 1
COMPARE
4000 500 Erlenmeyer Flask, Glass, 4000 mL, 1/pk
$18.00USD / Pkg of 1
COMPARE
5000 500 Erlenmeyer Flask, Glass, 5000 mL, 1/pk
$25.00USD / Pkg of 1
COMPARE
6000 500 Erlenmeyer Flask, Glass, 6000 mL, 1/pk
$27.50USD / Pkg of 1
Showing 1- 13 of 13