Cole-Parmer Ductless Fume Hood Filters
A filter for every application
  • Remove pollutants safety
Choose from a wide range of filters to remove a wide variety of pollutants. Filters are compatible with the Cole-Parmer range of ductless fume hoods
$185.00 - $734.00USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Filter Type Description
Availability Pricing
COMPARE
Carbon for organics and solvents Carbon Filter, Organics and Solvents
$305.00USD / Each
COMPARE
Carbon for acids and sulfur Carbon Filter, Acids and Sulfur
$372.00USD / Each
COMPARE
HEPA for powders and particulates HEPA Filter, Powders and Particulates
$377.00USD / Each
COMPARE
Carbon for formaldehyde Carbon Filter, Formaldehyde
$379.00USD / Each
COMPARE
Carbon for ammonia and amines Carbon Filter, Ammonia and Amines
$350.00USD / Each
COMPARE
Prefilter Accessory Prefilter; 6/Pk
$185.00USD / Pkg of 6
COMPARE
Carbon for organic vapors FHD-400 Ductless Fume Hood Carbon Filter for Organic Vapors including solvents and hydrocarbon vapors, units require qty 2 each.
$539.00USD / Each
COMPARE
Carbon for inorganic vapors FHD-400 Ductless Fume Hood Carbon Filter for Inorganic Vapors including acids, units required qty 2 each.
$637.00USD / Each
COMPARE
HEPA for powders and particulates FHD-400 Ductless Fume Hood HEPA Filter for Powders and Particulates, units require qty 2 each.
$734.00USD / Each
COMPARE
Carbon for lightweight aldehyde vapors FHD-400 Ductless Fume Hood Carbon Filter for Lightweight Aldehyde Vapors, such as formaldehyde and glutaraldehyde, units requires qty 2 each.
$658.00USD / Each
COMPARE
Carbon for organic vapors DDW-200 Downflow Workstation Carbon Filter for Organic Vapors including solvents and hydrocarbon vapors.
$549.00USD / Each
COMPARE
Carbon for inorganic vapors DDW-200 Downflow Workstation Carbon Filter for Inorganic Vapors including acids.
$641.00USD / Each
COMPARE
HEPA for powders and particulates DDW-200 Downflow Workstation HEPA Filter for powders and particulates.
$690.00USD / Each
COMPARE
Carbon for lightweight aldehyde vapors DDW-200 Downflow Workstation Carbon Filter for Lightweight Aldehyde Vapors, such as formaldehyde and glutaraldehyde.
$668.00USD / Each
Showing 1- 14 of 14