Cole-Parmer Dip, Stir, and Sample Kits
Versatile sampling kits with interchangeable parts
  • All components made of FDA-compliant HDPE
  • Purchase individual replacement components as needed!
All dipping beakers, stir blades, and sampling bottles use the same thread to be compatible with all three handle length options. Replacement handles and samplers may be replaced as needed — no need to purchase an entire kit if a part is lost or worn!
$370.00 - $541.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Sterile Description
Availability Pricing
COMPARE
HDPE No Complete Liquid Sampling and Stirring Kit
$541.00USD / Each
COMPARE
HDPE No Dipper Sampler Kit; HDPE
$370.00USD / Each