Cubitainer® Collapsible Polyethylene (PE) Containers
Collapsible containers nest for efficient storage
  • Meets Level 3 classification for contaminant-free sample containers
  • Just unscrew cap and fill to expand to total volume
Cubitainer® containers meet EPA recommended guidelines for contamination-free sample containers, but do not ship with a certificate of analysis.
$42.00 - $535.00USD / Case of 12
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Material Description
Availability Pricing
EPC3030 Cubitainer, LDPE, 1 Quart; 12/Cs EW-35204-79 Mfr # EPC3030
COMPARE
0.946 Low Density Polyethyene (LDPE) Cubitainer, LDPE, 1 Quart; 12/Cs
$48.50USD / Case of 12
EPC3020 Cubitainer, LDPE, 4 Liter; 12/Cs EW-35204-81 Mfr # EPC3020
COMPARE
4 Low Density Polyethyene (LDPE) Cubitainer, LDPE, 4 Liter; 12/Cs
$42.00USD / Case of 12
EPC3100 Cubitainer, LDPE, 4 Liter; 160/Cs EW-35204-83 Mfr # EPC3100
COMPARE
4 LDPE Cubitainer, LDPE, 4 Liter; 160/Cs
$495.00USD / Case of 160
EPC3010 Cubitainer, LDPE, 10 Liter; 36/Cs EW-35204-84 Mfr # EPC3010
COMPARE
10 Low Density Polyethyene (LDPE) Cubitainer, LDPE, 10 Liter; 36/Cs
$173.00USD / Case of 36
EPC3000 Cubitainer, LDPE, 20 Liter; 36/Cs EW-35204-88 Mfr # EPC3000
COMPARE
20 Low Density Polyethyene (LDPE) Cubitainer, LDPE, 20 Liter; 36/Cs
$285.00USD / Case of 36
EPC3155 Cubitainer, LDPE, 1 Liter; 128/Cs EW-35204-90 Mfr # EPC3155
COMPARE
1 LDPE Cubitainer, LDPE, 1 Liter; 128/Cs
$535.00USD / Case of 128
COMPARE
4 High Density Polyethyene (HDPE) Cubitainer, HDPE, 4 Liter; 160/Cs
$529.50USD / Case of 160
COMPARE
1 High Density Polyethyene (HDPE) Cubitainer, HDPE, 1 Liter; 128/Cs
$443.00USD / Case of 128
Showing 1- 8 of 8