Cole-Parmer Cryogenic Labels
  • Perfect for cryogenic storage — withstand temperatures from -321 to 302°F (-196 to 150°C).
These labels will withstand conventional and cryogenic storage to -321°F (-196°C), boiling water baths to 212°F (100°C), and dry heat up to 302°F (150°C). Labels adhere to most plastics, glass, and metals. Resistant to detergents, oils, solvents, and caustic agents without peeling or falling off.
$68.70USD / Roll of 1000
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Temperature Points (° C) Width (in)
Availability Pricing
COMPARE
1/2 -196 to 150 1 9/32
$68.70USD / Roll of 1000
COMPARE
3/4 -196 to 150 1 1/2
$68.70USD / Roll of 1000