Cole-Parmer® Contact Tachometers with LCD/LED Display
Rugged aluminum construction – ideal for industrial applications
  • Easily switch between 16 measuring modes with front-panel dial
Dial in any one of the 16 measuring modes — these tachometers calculate the readings for you! Select the LCD model for use in direct sunlight, or choose the LED model for easy reading in dim lighting. Both models feature a large memory capacity — use the memory button to recall maximum, minimum, and the last ten readings. Low-battery indicator notifies user of battery life.
$270.00 - $355.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Contact Tachometer with LCD Display
$270.00USD / Each
COMPARE
Contact Tachometer with LED Display
$354.00USD / Each
COMPARE
Contact Tachometer with LCD Display and 12\" (30.5 cm) Wheel
$355.00USD / Each