Cole-Parmer® Compact Fume Hoods
Economical and efficient system for your basic fume hood needs
  • Complete with dished composite resin work surface, vaporproof light, and built-in exhaust blower.
Fume chamber features molded seamless construction with coved corners, factory mounted with a chemical-resistant composite resin work surface to contain spills. Varaflow baffle system is designed to efficiently direct air through the chamber. Viewing sash is constructed of 3⁄16"-thick tempered safety glass with chemical-resistant PVC frame and track. Vaporproof light and exhaust blower, with polypropylene impeller, are built in with columnmounted switches. Blower rated for 12-ft maximum duct run.
Please note: cabinet units do not come with exhaust blowers. Units do not come with power cords and must be hard-wired.
$14,048.00 - $16,010.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Power (VAC) Description
Availability Pricing
COMPARE
45 115 Fume Hood, 30\", Dish Work Surface/VP Light/Blower; 115 V
$14,335.00USD / Each
COMPARE
45 115 Fume Hood, 36\", Dish Work Surface/VP Light/Blower; 115 V
$14,048.00USD / Each
COMPARE
45 115 Fume Hood, 48\", Dish Work Surface/VP Light/Blower; 115 V
$16,010.00USD / Each