Cole-Parmer® Cleanroom Vinyl Ergonomic Chair

Class 100/ISO Class 5 cleanroom chairs are comfortable and easy to clean
  • Heavy-duty vinyl seat features molded polyurethane foam cushioning
  • Ergonomic seat tilts 8° forward, 3° backward, and locks in place
  • Rugged and durable weight capacity of 300 lbs.
These Class 100/ISO Class 5 cleanroom chairs are made of molded polyurethane foam cushioning covered with easily cleaned antibacterial/antimicrobial vinyl. Ergonomic seat tilts forward, backward, and locks in place for ergonomic posture support. The contoured waterfall seat cushion and an adjustable back rest with lumbar support for increased comfort during extended sitting durations.

Cleanroom chairs have been tested to Federal Standard 209E and ISO 14644-1 (Class 100/ISO Class 5) standards.

All chairs feature a smooth pneumatic lift. Choose from desk or bench height chairs with pneumatic adjustment. Chairs are also available in black or blue colors, with or without adjustable arm rest and foot ring. Choose casters or glides, both of which are non-marring and suitable for hard or carpeted floors.
$454.00 - $643.00USD / Each

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Material Description
Availability Pricing
COMPARE
Black Vinyl Clean Room Ergonomic Chair, Vinyl, Desk Height, Black, No Arms, Chrome Casters
$454.00USD / Each
COMPARE
Black Vinyl Cleanroom Ergonomic Chair, Vinyl, Desk Height, Black, Adjustable Arms, Chrome Casters
$579.00USD / Each
COMPARE
Blue Vinyl Cleanroom Ergonomic Chair, Vinyl, Desk Height, Blue, Adjustable Arms, Chrome Casters
$579.00USD / Each
COMPARE
Black Vinyl Cleanroom Ergonomic Chair, Vinyl, Desk Height, Black, Adjustable Arms, Gliders
$579.00USD / Each
COMPARE
Blue Vinyl Cleanroom Ergonomic Chair, Vinyl, Desk Height, Blue, Adjustable Arms, Gliders
$579.00USD / Each
COMPARE
Black Vinyl Cleanroom Ergonomic Chair, Vinyl, Bench Height, Black, No Arms, Chrome Casters, Chrome Foot Ring
$519.00USD / Each
COMPARE
Blue Vinyl Cleanroom Ergonomic Chair, Vinyl, Bench Height, Blue, No Arms, Chrome Casters, Chrome Foot Ring
$519.00USD / Each
COMPARE
Black Vinyl Cleanroom Ergonomic Chair, Vinyl, Bench Height, Black, No Arms, Gliders, Chrome Foot Ring
$519.00USD / Each
COMPARE
Black Vinyl Cleanroom Ergonomic Chair, Vinyl, Bench Height, Black, No Arms, Gliders, No Foot Ring
$473.00USD / Each
COMPARE
Black Vinyl Cleanroom Ergonomic Chair, Vinyl, Bench Height, Black, Adjustable Arms, Chrome Casters, Chrome Foot Ring
$643.00USD / Each
COMPARE
Blue Vinyl Cleanroom Ergonomic Chair, Vinyl, Bench Height, Blue, Adjustable Arms, Chrome Casters, Chrome Foot Ring
$643.00USD / Each
COMPARE
Black Vinyl Cleanroom Ergonomic Chair, Vinyl, Bench Height, Black, Adjustable Arms, Chrome Casters, No Foot Ring
$598.00USD / Each
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat