Cole-Parmer Barometric Pressure Transducers
Excellent response time and long-term stability
  • Sleep mode ensures instant start-up
  • Low power consumption — 3 mA nominal
Low-cost barometric pressure transducers are designed for use in environmental applications that require excellent accuracy, fast response, and long term reliability. Accurate signal conditioning and environmental compensation are provided by a capacitive sensor and proprietary custom IC circuit.
Housing is constructed of stainless steel and polyester that withstand environmental extremes. A removable 5-pin terminal strip enables easy wiring connection to datalogger and signal connections. Pressure connection is a 1/8" barbed fitting.
ITEM(S) ON SALE
$963.05USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
COMPARE
±0.3 hPa/mb 0.01 mb
$963.05USD / Each
$1,133.00USD / Each