Cole-Parmer® Essentials Amber-Scale Graduated Glass Cylinders
Accurate measurements even at lowest graduations
  • Calibrated “To Deliver”
  • Single metric scale
Dark amber graduations make these cylinders ideal for use with light colored liquids. Cylinders provide accurate measurements even at the lowest graduations; the first 10% of volume is not graduated. Designed from ASTM E 1272, Style 1, Class B requirements. Cylinders feature heavy round bases and convenient pouring spouts.
$31.00 - $158.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
5 0.1 Glass
$31.00USD / Each
COMPARE
10 0.2 Glass
$31.00USD / Each
COMPARE
50 1 Glass
$39.00USD / Each
COMPARE
250 2 Glass
$72.00USD / Each
COMPARE
500 5 Glass
$95.00USD / Each
COMPARE
1000 10 Glass
$110.00USD / Each
COMPARE
2000 20 Glass
$158.00USD / Each
Showing 1- 7 of 7