Cole-Parmer® Adjustable-Angle Industrial Silicone-Filled Bimetal Thermometers
Silicone filled to provide superior response times and greatly reduce pointer flutter
  • Hermetically sealed case prevents entry of moisture, interior corrosion and coil freeze up
  • Anti-parallax dial for precise readings of both scales
  • External adjustment reduces maintenance
  • NEMA 4X/IP66 protection
  • Able to perform quick and limited span adjustments
  • All-welded stainless steel construction
Silicone-filled bimetal thermometers are best suited to high-vibration applications. Available in multiple temperature ranges and dial sizes for a variety of needs.
$163.00 - $196.00USD / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Dial Size (in) Max Temperature (° C) Min Temperature (° C)
Availability Pricing
COMPARE
3 120 -20
$163.00USD / Each
COMPARE
3 120 -20
$163.00USD / Each
COMPARE
3 120 -20
$163.00USD / Each
COMPARE
3 120 -20
$163.00USD / Each
COMPARE
3 120 -20
$163.00USD / Each
COMPARE
3 290 10
$163.00USD / Each
COMPARE
3 290 10
$163.00USD / Each
COMPARE
3 290 10
$163.00USD / Each
COMPARE
3 290 10
$163.00USD / Each
COMPARE
3 290 10
$163.00USD / Each
COMPARE
5 120 -20
$196.00USD / Each
COMPARE
5 120 -20
$196.00USD / Each
COMPARE
5 120 -20
$196.00USD / Each
COMPARE
5 120 -20
$196.00USD / Each
COMPARE
5 120 -20
$196.00USD / Each
COMPARE
5 290 10
$196.00USD / Each
COMPARE
5 290 10
$196.00USD / Each
COMPARE
5 290 10
$196.00USD / Each
COMPARE
5 290 10
$196.00USD / Each
COMPARE
5 290 10
$196.00USD / Each
Showing 1- 20 of 20