Class A Volumetric Pipettes
  • Reusable, Class A borosilicate glass pipettes meet ASTM requirements
Volume accuracies and capacities are permanently marked in amber on each pipette. Pippettes are calibrated "to deliver". All pipettes feature treated and tempered tips to reduce chipping and breakage. Class A Pipettes meet all the requirements of ASTM E 969-95. Compliance with the CAP and other laboratory certifying groups is indicated by the letter “A” near the top of each pipette. All pipettes are calibrated in accordance with ASTM E 542 and meet the accuracy requirements of ASTM E 969; borosilicate glass meets ASTM E 438 for Type I, Class A requirements.
Class B Pipettes meet the flow time and volume accuracy requirements of ASTM E 969-95 and are marked with a “B” near the top of each pipette.
$40.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Volume (mL)
Availability Pricing
DWK Life Sciences (Kimble) Volumetric Pipettes, Class A pipettes, 25 mL EW-25560-26 Mfr # CP8170AP
COMPARE
25
$40.00USD / Each