This product is now discontinued, View Related Products
Chemical Splash Suits 7693
Best value in chemical splash protection
Splash suits consist of coveralls with an attached hood made from 0.35-mm heavy-duty PVC on nylon/polyester. Features include reinforced crotch, elbows, and knees; heavy-duty elastic suspender with clip; adjustable waist snaps; heavy-duty zipper; cord locks on hood strings; and ankle and cuff take-up snaps for a comfortable fit. Wallet size degradation chart is included. For use in Level C environments. Compliance: Meets Federal Apparel Flammability Act, FDS 5903.1
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Color Size
Availability Pricing
Onguard 71022-M Chemical Splash Suit, Hooded, Medium; 1/Pack EW-86234-01 Mfr # 71022-M
COMPARE
Green Medium
Call For Price
Onguard 71022-L Chemical Splash Suit, Hooded,Large; 1/Pack EW-86234-02 Mfr # 71022-L
COMPARE
Green Large
Call For Price
Onguard 71022- Chemical Splash Suit, Hooded, X-Large; 1/Pack EW-86234-03 Mfr # 71022-XL
COMPARE
Green X-Large