Charts for Barton, American Meter, and Mercury Recorders
Get exacting performance that meets American Gas Association (AGA) standards
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
ITEM(S) ON SALE
$52.50 - $74.20USD / Box of 100
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chart Diameter (in) Chart Speed Max Input (VDC)
Availability Pricing
Graphic Controls 00002766 Circular Chart, 7 day, 12", 0-10 scale EW-49000-42 Mfr # 00002766
COMPARE
12 7 day
$52.50USD / Box of 100
$75.00USD / Box of 100
Graphic Controls 00006932 Circular Chart, 7 day, 12" dia, 0-100 scale (x2) EW-49000-45 Mfr # 00006932
COMPARE
12 7 day
$52.50USD / Box of 100
$75.00USD / Box of 100
Graphic Controls 00015982 Circular Chart, 7 day, 12" dia, 0-100 & 0-500 scales EW-49000-47 Mfr # 00015982
COMPARE
12 7 day
$52.50USD / Box of 100
$75.00USD / Box of 100
Graphic Controls 30269057 Circular Chart, 24 hour EW-49000-49 Mfr # 30269057
COMPARE
12 1 day
$56.00USD / Box of 100
$80.00USD / Box of 100
Graphic Controls 00009076 Circular Chart, 7 day, 12" dia, 0-100 & 0-1000 scales EW-49000-50 Mfr # 00009076
COMPARE
12 7 day
$52.50USD / Box of 100
$75.00USD / Box of 100
Graphic Controls 00010694 Circular Chart, American Meter/Barton, 7 day EW-49000-51 Mfr # 00010694
COMPARE
12 7 day
$52.50USD / Box of 100
$75.00USD / Box of 100
Graphic Controls 31232427 Circular Chart, American Meter/Barton, 24 hour EW-49000-55 Mfr # 31232427
COMPARE
12 1 day
$52.50USD / Box of 100
$75.00USD / Box of 100
Graphic Controls 31504239 Circular Chart, American Meter/Barton, 1 hour EW-49000-56 Mfr # 31504239
COMPARE
12 1 hour
$52.50USD / Box of 100
$75.00USD / Box of 100
Graphic Controls 00787960 Circular Chart, American Meter/Barton, 24 hour EW-49000-58 Mfr # 00787960
COMPARE
12 1 day
$52.50USD / Box of 100
$75.00USD / Box of 100
Graphic Controls 00983627 Circular Chart; American Meter/Barton, 8 day EW-49000-60 Mfr # 00983627
COMPARE
12 8 day
$52.50USD / Box of 100
$75.00USD / Box of 100
Graphic Controls 00380923 Circular Chart, Mercury Instrument, 31 day EW-49000-61 Mfr # 00380923
COMPARE
8 31 day
$74.20USD / Box of 100
$106.00USD / Box of 100
Graphic Controls 00378661 Circular Chart, Mercury Instrument, 7 day EW-49000-62 Mfr # 00378661
COMPARE
8 7 day
$69.30USD / Box of 100
$99.00USD / Box of 100
Showing 1- 12 of 12