Centrifuges for Mojonnier Ether Extraction
Use these centrifuges to perform acid hydrolysis for your fat analysis. Small centrifuge 17360-40 holds up to 8 Mojonnier flasks; large centrifuge 17360-45 holds up to 12 Mojonnier flasks.
Centrifuges are constructed of 319 aluminum and polished to a satin finish. Both models feature sealed manual brakes and safety switch. Easily mount external motor switch up to 6-ft away. Switch is enclosed in an explosion-proof and waterproof housing.
$12,070.00 - $12,966.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RCF x g Max Speed (rpm) Description
Availability Pricing
Mojonnier 110V, centrifuge; 8 bottle capacity EW-17360-01 Mfr # 308M - 115V/60HZ
COMPARE
100 600 Mojonnier 110V, centrifuge; 8 bottle capacity
$12,070.00USD / Each
Mojonnier 220V, centrifuge; 12 bottle capacity EW-17360-45 Mfr # 312M - 220V/50HZ
COMPARE
100 600 Mojonnier 220V, centrifuge; 12 bottle capacity
$12,966.00USD / Each