CELLTREAT® Cell Scrapers
Easily harvest cells from tissue culture flasks, dishes, or bottles
  • Soft, pliable blades gently remove cells and maximize recovery
Scrapers feature ABS handle and TPE blade. Certified nonpyrogenic and packaged sterile (gamma irradiated) in individual wrappers.
$183.75 - $222.25USD / Case of 100
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Overall Length (in) Sterile Description
Availability Pricing
CELLTREAT Scientific Products 229310 Cell Scraper, 25 cm Handle, 20 mm “Scraper” Blade, Sterile; 100/Cs EW-01959-30 Mfr # 229310
COMPARE
Yes Cell Scraper, 25 cm Handle, 20 mm “Scraper” Blade, Sterile; 100/Cs
$187.75USD / Case of 100
CELLTREAT Scientific Products 229311 Cell Scraper, 25 cm Handle, 20 mm “Lifter” Blade, Sterile; 100/Cs EW-01959-32 Mfr # 229311
COMPARE
Yes Cell Scraper, 25 cm Handle, 20 mm “Lifter” Blade, Sterile; 100/Cs
$183.75USD / Case of 100
CELLTREAT Scientific Products 229312 Cell Scraper, 30 cm Handle, 20 mm "Pivoting" Blade, Sterile; 100/Cs EW-01959-34 Mfr # 229312
COMPARE
Yes Cell Scraper, 30 cm Handle, 20 mm \"Pivoting\" Blade, Sterile; 100/Cs
$222.25USD / Case of 100
CELLTREAT Scientific Products 229315 Cell Scraper, 39 cm Handle, 30 mm “Scraper” Blade, Sterile; 100/Cs EW-01959-36 Mfr # 229315
COMPARE
Yes Cell Scraper, 39 cm Handle, 30 mm “Scraper” Blade, Sterile; 100/Cs
$211.25USD / Case of 100
CELLTREAT Scientific Products 229316 Cell Scraper, 39 cm Handle, 30 mm “Lifter” Blade, Sterile; 100/Cs EW-01959-38 Mfr # 229316
COMPARE
Yes Cell Scraper, 39 cm Handle, 30 mm “Lifter” Blade, Sterile; 100/Cs
$213.75USD / Case of 100