This product is now discontinued, View Related Products
J.H. Williams Tool Group Dial Torque Wrenches
Memory needle acts as an indicator of how much torque has been applied
  • Exclusive bi-polar dual scales, English and Metric, permits accurate reading of both scales Equally accurate in both left and right hand direction Torsion drive design Large easy to read dial User assured of long life and reliability Calibration traceable to National Institute of Standards and Technology (NIST) Available with Follower Needle, Electric Light and Beeper or Ratcheting Drive (LDFNR) Special dual scale combinations (torque ranges & configurations) Includes plastic case
The pointer needle returns to zero but the memory needle remains at the highest applied torque value. Electric Dial (Alarm) Operator can preset a desired torque value. When the pointer needle reaches the preset torque value, the light is illuminated and a buzzer sounds.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Capacity (lb-in) Capacity (N-m)
Availability Pricing
J. H. Williams Tool Group 2502LDIN Torque Wrench 3-8in 0/250 Inlbs.W/Fllwer Needle Dual Scale EW-97150-16 Mfr # 2502LDIN
COMPARE
Call For Price
J. H. Williams Tool Group 3002LDIN Dial Indicating Torque Wrench, 3/8" Drive, 0-300 in lb EW-97150-17 Mfr # 3002LDIN
COMPARE
300 35
Call For Price
J. H. Williams Tool Group 301LDIN Dial Indicating Torque Wrench, 1/4" Drive, 0-30 in lb EW-97150-18 Mfr # 301LDIN
COMPARE
30 3.5
Call For Price
J. H. Williams Tool Group 151LDIN Torque Wrench 1-4in 0/15 Inlbs.W/Fllower Needle Dual Scale EW-97150-24 Mfr # 151LDIN
COMPARE
Call For Price
J. H. Williams Tool Group 6002LDIN Dial Indicating Torque Wrench, 3/8" Drive, 0-600 in lb EW-97150-25 Mfr # 6002LDIN
COMPARE
600 70
Call For Price
J. H. Williams Tool Group 1502LDIN Torque Wrench 3-8in 0/150 Inlbs.W/Fllwer Needle Dual Scale EW-97150-27 Mfr # 1502LDIN
COMPARE
Call For Price
J. H. Williams Tool Group 751LDIN Dial Indicating Torque Wrench, 1/4" Drive, 0-75 in lb EW-97150-30 Mfr # 751LDIN
COMPARE
75 9
Call For Price
Showing 1- 7 of 7