Calibrated Replacement Probes for Traceable® Data Logging Ethernet Thermometers
Save time and money by simply replacing probes that are damaged or due for calibration
  • No need to send in your thermometer for re-calibration—simply replace the recalibrated probe
  • Get accurate, reliable measurements with included NIST-traceable calibration
These replacement temperature probes are compatible with the Traceable® Data Logging Ethernet Thermometers.
Traceable® products are calibrated to NIST-traceable standards and ready to use as soon as you receive it. Each product goes through an intense vetting process before it can carry the Traceable name, and it includes NIST-traceable certification. This means that a 17025-accredited calibration test lab has confirmed that the product meets or exceeds all stated specifications, saving you both time and money by not having to calibrate separately.
NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.
$108.00 - $156.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (in) Material Compatible With
Availability Pricing
COMPARE
120 Stainless Steel 18004-37
$156.00USD / Each
COMPARE
120 Glycol-filled bottle probe 99460-01
$108.00USD / Each