This product is now discontinued, View Related Products
Burkle Disposable Multiple-Layer Sampling Lances for Powders and Small Granulates
Quickly empty samples through open handle design
  • Openings along shaft provide multiple-layer sampling
  • Idea for free-flowing powders and small granulates
  • Gamma sterilized and nonsterile versions available
  • Manufactured in cleanroom environment and individually packaged
These disposable HDPE samplers are ideal for collecting multiple-layer powder and small granulated samples from sacks or bags. A specially designed opening in the handle allows your all-layer sample to be directly deposited into a container — no disassembly required. Choose from either nonsterile or sterile configurations; the sterile option is sterilized via gamma irradiation.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Sterile Volume (mL)
Availability Pricing
Burkle 5393-4440 Disposable Multiple-Layer Sampling Lance, HDPE, 50 cm length; 100 mL EW-06043-19 Mfr # 5393-4440
COMPARE
19.7 No 100
Call For Price
Burkle 5393-4441 Disposable Multiple-Layer Sampling Lance, HDPE, Sterile, 50 cm length; 100 mL EW-06043-21 Mfr # 5393-4441
COMPARE
19.7 Yes 100
Call For Price
Burkle 5393-4450 Disposable Multiple-Layer Sampling Lance, HDPE, 100 cm length; 200 mL EW-06043-22 Mfr # 5393-4450
COMPARE
39.4 No 200
Call For Price
Burkle 5393-4451 Disposable Mulitple-Layer Sampling Lance, HDPE, Sterile, 100 cm length; 200 mL EW-06043-23 Mfr # 5393-4451
COMPARE
39.4 Yes 200