Bud Industries Sealed Polycarbonate and ABS Enclosures
Ideal for PCB enclosures, junction boxes and other applications in electrical, process, and control industries
  • Designed to IP65 of IEC 529 and NEMA 4X specifications
  • Wall mounting holes and cover attaching screws are outside the sealing area thus preventing ingress of moisture and dust
  • Cover screws are M-4 stainless steel, non-magnetic and fasten into threaded brass inserts
These Polycarbonate and high-impact ABS molded enclosures meet NEMA 4X specifications and are. Available in a variety of sizes, these molded enclosures are offered in. Light gray polycarbonate body and clear polycarbonate cover. Body/Cover - UV stabilized , UL94V-2 flammability rating, suggested service temperature -40 to 120°C. PN-Series enclosures feature a smooth, flat top cover. In order to maintain NEMA 4X rating, cable glands must be used when cables enter or exit the enclosure.
$10.80 - $19.30USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
Bud Industries PN-1320 Enclosure with light gray polycarbonate body and cover; 2.28" x 2.56" x 1.38" EW-94501-18 Mfr # PN-1320
COMPARE
1.38 2.56 2.28
$10.80USD / Each
Bud Industries PN-1321 Enclosure with light gray polycarbonate body and cover; 2.56" x 4.53" x 1.57" EW-94501-20 Mfr # PN-1321
COMPARE
1.57 4.53 2.56
$15.60USD / Each
Bud Industries PN-1323 Enclosure with light gray polycarbonate body and cover; 3.54" x 4.53" x 2.17" EW-94501-26 Mfr # PN-1323
COMPARE
2.17 4.53 3.54
$19.30USD / Each
Bud Industries PN-1327-DG Enclosure with dark gray ABS body and cover; 4.76" x 6.73" x 3.15" EW-94501-32 Mfr # PN-1327-DG
COMPARE
3.15 6.73 4.76
$17.70USD / Each