Bud Industries NBA-Series Instrument and Control Enclosures
Clear, hinged doors enable easy viewing and quick access to controls and meters
  • For indoor use
  • Enclosures meet NEMA-4X and IEC529-IP66 requirements
  • Clear doors are polycarbonate plastic with a UL94V-0 flammability rating
These enclosures are perfect for numerous sizes of controls, instruments, meters, or gauges, as well as electronics or electrical devices. Grey in color, enclosures are molded out of PC/PBT blended plastic. Clear doors are gasketed, and have stainless steel hinges and latches. Internal mounting bosses enable user to securely attached DIN rails or mounting brackets that secure instruments and controls.
$235.25 - $280.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
Bud Industries NBA-10168 Enclosure, ABS, 7" x 12" x 16" EW-94503-04 Mfr # NBA-10168
COMPARE
7.09 15.75 11.8
$235.25USD / Each
Bud Industries NBA-10172 Instrument Enclosure, ABS, 8" X 16" X 20" EW-94503-06 Mfr # NBA-10172
COMPARE
7.87 19.68 15.75
$280.00USD / Each