BTX Electroporation Cuvettes, Sterile
High quality cuvettes for transfection/transformation of bacteria, yeast, insect, plant and mammalian cells
  • Compatible with most electroporation systems
  • Available in 1mm, 2mm and 4mm gap sizes
  • Color-coded for easy identification
  • Round caps for ease of use
  • Sterile cuvettes with a disposable pipette
BTX Cuvettes Plus are designed for use in electroporation and electrofusion of bacteria, yeast, insect, plant and mammalian cells. Each sterilized Cuvettes Plus package includes a disposable cuvette and a bonus pipette, which is used for removal of the sample after electroporation. The cuvettes are molded with embedded aluminum electrodes, washed prior to packaging and gamma irradiated for guaranteed sterility. Three electrode gap sizes are available, 4mm, 2mm and 1mm for high field strengths up to 25.0 kV/cm. Round caps are leak resistant and allow for quick and easy one finger removal. BTX disposable Cuvettes Plus require no maintenance. Simply discard carefully after use. For research use only.
$398.00 - $532.00USD / Pkg of 50
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Capacity (MicroL) Description
Availability Pricing
BTX 45-0124 Electroporation Cuvettes, Sterile, Gray, 1 mm Gap; 50/Pk EW-02894-30 Mfr # 45-0124
COMPARE
90 Electroporation Cuvettes, Sterile, Gray, 1 mm Gap; 50/Pk
$398.00USD / Pkg of 50
BTX 45-0125 Electroporation Cuvettes, Sterile, Blue, 2 mm Gap; 50/Pk EW-02894-32 Mfr # 45-0125
COMPARE
400 Electroporation Cuvettes, Sterile, Blue, 2 mm Gap; 50/Pk
$532.00USD / Pkg of 50
BTX 45-0126 Electroporation Cuvettes, Sterile, Yellow, 4 mm Gap; 50/Pk EW-02894-34 Mfr # 45-0126
COMPARE
800 Electroporation Cuvettes, Sterile, Yellow, 4 mm Gap; 50/Pk
$448.00USD / Pkg of 50