Ready-to-use heating cord bundles deliver complete systems for exceptional convenience
Programmable controller features alarm, user selectable °C or °F, and advanced PID control
Easy to read measurements with 0.4” high, 4-digit, green LED display
Constructed of heavy insulated knitted and braided fiberglass with silicone rubber outer sheath
Maximum temperature of 232°C (450°F)
All-inclusive bundle features a digital PID controller with silicone rubber heating tape, and power cords—everything you need to get started. Benchtop controller is compact and easy to program, featuring a plug-and-play design. The X2 series controller offers a simple three button control with a programmable temperature, alarms, and security locks.
The flexible silicone rubber heating tape is moisture, chemical, and radiation resistant to provide ideal heating for flat and curved surface heating. The heating tape has a minimum bend radius of 0.25” (6 mm) with a nominal thickness of 0.125” (3 mm).
The heavy insulated heating tapes feature an IP66 rating, making them ideal for rugged applications. Select heating tape length to optimize coverage while avoiding overlap.