BriskHeat® Silicone-Rubber Heater Blankets Metal Surfaces - 120 VAC
Durable heater is ideal for uniform heating of large surfaces such as tanks and hoppers
  • Maximum exposure temperature of 450°F (232°C)
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications. The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Heaters for metal surfaces have faster heat-up rates due to the 2.5 watts /in2 power density.
$118.00 - $638.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRL06121 Silicone Heating Blanket, 6x12 Size, 120 Volt, 180 Watt, for metal surfaces EW-36001-26 Mfr # SRL06121
COMPARE
12 450 120
$118.00USD / Each
BriskHeat SRL06241 Silicone Heating Blanket, 6x24 Size, 120 Volt, 360 Watt, for metal surfaces EW-36001-28 Mfr # SRL06241
COMPARE
24 450 120
$169.00USD / Each
BriskHeat SRL06361 Silicone Heating Blanket, 6x36 Size, 120 Volt, 540 Watt, for metal surfaces EW-36001-31 Mfr # SRL06361
COMPARE
36 450 120
$233.00USD / Each
BriskHeat SRL12121 Silicone Heating Blanket, 12x12 Size, 120 Volt, 360 Watt, for metal surfaces EW-36001-33 Mfr # SRL12121
COMPARE
12 450 120
$155.00USD / Each
BriskHeat SRL12241 Silicone Heating Blanket, 12x24 Size, 120 Volt, 720 Watt, for metal surfaces EW-36001-36 Mfr # SRL12241
COMPARE
24 450 120
$264.00USD / Each
BriskHeat SRL12361 Silicone Heating Blanket, 12x36 Size, 120 Volt, 1080 Watt, for metal surfaces EW-36001-38 Mfr # SRL12361
COMPARE
36 450 120
$420.00USD / Each
BriskHeat SRL18181 Silicone Heating Blanket, 18x18 Size, 120 Volt, 810 Watt, for metal surfaces EW-36001-41 Mfr # SRL18181
COMPARE
18 450 120
$323.00USD / Each
BriskHeat SRL18361 Silicone Heating Blanket, 18x36 Size, 120 Volt, 1620 Watt, for metal surfaces EW-36001-43 Mfr # SRL18361
COMPARE
36 450 120
$622.00USD / Each
BriskHeat SRL24241 Silicone Heating Blanket, 24x24 Size, 120 Volt, 1440 Watt, for metal surfaces EW-36001-48 Mfr # SRL24241
COMPARE
24 450 120
$486.00USD / Each
BriskHeat SRL24361 Silicone Heating Blanket, 12x36 Size, 120 Volt, 2160 Watt, for metal surfaces EW-36001-50 Mfr # SRL24361
COMPARE
36 450 120
$638.00USD / Each
Showing 1- 10 of 10