BriskHeat® Fiberglass Heating Cords with SDC Digital Temperature Controller Bundles
Convenient bundle with your choice of heating cord
  • All-inclusive, plug-and-play benchtop temperature controller has everything you need
  • Easy to program and operate
  • Cords have exceptional flexibility; can be wrapped around objects as small as 1/8" (3 mm) diameter
This bundle starts with your choice of a digital temperature controller that displays its temperature reading in Fahrenheit or Celsius. It has a temperature control range of 32 to 999 °F (0 to 700 °C) with an accuracy rate of +/- 1% full-scale. Includes a type J thermocouple (type K thermocouple is also available; call for details). Dimensions: 5.5" L x 4.25" W x 1.75" H (14.0 x 10.8 x 4.5 cm).
Next, choose a standard or high temperature heating cord. They are constructed of a double-braided fiberglass outer sheath with a diameter of 3/16"(4.8 mm).
$412.00 - $481.00USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Power (watts) Description
Availability Pricing
BriskHeat SDCJFA-HTC451005 Heating Cord with SDC Temp Controller, 2 ft (0.6 m) Length, 43 Watts, °F Temp Display; 120 VAC EW-36120-11 Mfr # SDCJFA-HTC451005
COMPARE
900 43 Heating Cord with SDC Temp Controller, 2 ft (0.6 m) Length, 43 Watts, °F Temp Display; 120 VAC
$413.00USD / Each
BriskHeat SDCJFA-HTC451001 Heating Cord with SDC Temp Controller, 3 ft (0.9 m) Length, 64 Watts, °F Temp Display; 120 VAC EW-36120-12 Mfr # SDCJFA-HTC451001
COMPARE
900 64 Heating Cord with SDC Temp Controller, 3 ft (0.9 m) Length, 64 Watts, °F Temp Display; 120 VAC
$415.00USD / Each
BriskHeat SDCJFA-HTC451008 Heating Cord with SDC Temp Controller, 4 ft (1.2 m) Length, 82 Watts, °F Temp Display; 120 VAC EW-36120-13 Mfr # SDCJFA-HTC451008
COMPARE
900 82 Heating Cord with SDC Temp Controller, 4 ft (1.2 m) Length, 82 Watts, °F Temp Display; 120 VAC
$419.00USD / Each
BriskHeat SDCJFA-HTC451003 Heating Cord with SDC Temp Controller, 12 ft (3.7 m) Length, 260 Watts, °F Temp Display; 120 VAC EW-36120-17 Mfr # SDCJFA-HTC451003
COMPARE
900 260 Heating Cord with SDC Temp Controller, 12 ft (3.7 m) Length, 260 Watts, °F Temp Display; 120 VAC
$452.00USD / Each
BriskHeat SDCJCA-HTC451007 Heating Cord with SDC Temperature Controller, 0.5 ft (0.15 m) Length, 17 Watts,°C Temp Display; 120 VAC EW-36120-18 Mfr # SDCJCA-HTC451007
COMPARE
900 17 Heating Cord with SDC Temperature Controller, 0.5 ft (0.15 m) Length, 17 Watts,°C Temp Display; 120 VAC
$412.00USD / Each
BriskHeat SDCJCA-HTC451006 Heating Cord with SDC Temp Controller, 1 ft (0.3 m) Length, 22 Watts, °C Temp Display; 120 VAC EW-36120-19 Mfr # SDCJCA-HTC451006
COMPARE
900 22 Heating Cord with SDC Temp Controller, 1 ft (0.3 m) Length, 22 Watts, °C Temp Display; 120 VAC
$412.00USD / Each
BriskHeat SDCJCA-HTC451005 Heating Cord with SDC Temp Controller, 2 ft (0.6 m) Length, 43 Watts, °C Temp Display; 120 VAC EW-36120-21 Mfr # SDCJCA-HTC451005
COMPARE
900 43 Heating Cord with SDC Temp Controller, 2 ft (0.6 m) Length, 43 Watts, °C Temp Display; 120 VAC
$413.00USD / Each
BriskHeat SDCJCA-HTC451008 Heating Cord with SDC Temp Controller, 4 ft (1.2 m) Length, 82 Watts, °C Temp Display; 120 VAC EW-36120-23 Mfr # SDCJCA-HTC451008
COMPARE
900 82 Heating Cord with SDC Temp Controller, 4 ft (1.2 m) Length, 82 Watts, °C Temp Display; 120 VAC
$419.00USD / Each
BriskHeat SDCJCA-HTC451009 Heating Cord with SDC Temp Controller, 8 ft (2.4 m) Length, 170 Watts, °C Temp Display; 120 VAC EW-36120-26 Mfr # SDCJCA-HTC451009
COMPARE
900 170 Heating Cord with SDC Temp Controller, 8 ft (2.4 m) Length, 170 Watts, °C Temp Display; 120 VAC
$437.00USD / Each
BriskHeat SDCJCA-HTC451003 Heating Cord with SDC Temp Controller, 12 ft (3.7 m) Length, 260 Watts, °C  Temp Display; 120 VAC EW-36120-27 Mfr # SDCJCA-HTC451003
COMPARE
900 260 Heating Cord with SDC Temp Controller, 12 ft (3.7 m) Length, 260 Watts, °C Temp Display; 120 VAC
$452.00USD / Each
BriskHeat SDCJFA-HWC1060 High Temp Heating Cord with SDC Temp Controller, 6 ft (1.8 m) Length, 350 Watts, °F Temp Display; 120 VAC EW-36120-29 Mfr # SDCJFA-HWC1060
COMPARE
999 350 High Temp Heating Cord with SDC Temp Controller, 6 ft (1.8 m) Length, 350 Watts, °F Temp Display; 120 VAC
$458.00USD / Each
BriskHeat SDCJFA-HWC1180 High Temp Heating Cord with SDC Temp Controller, 18 ft (5.4 m) Length, 1080 Watts, °F Temp Display; 120 VAC EW-36120-32 Mfr # SDCJFA-HWC1180
COMPARE
999 1080 High Temp Heating Cord with SDC Temp Controller, 18 ft (5.4 m) Length, 1080 Watts, °F Temp Display; 120 VAC
$481.00USD / Each
BriskHeat SDCJCA-HWC1040 High Temp Heating Cord with SDC Temp Controller, 4 ft (1.2 m) Length, 245 Watts, °C Temp Display; 120 VAC EW-36120-33 Mfr # SDCJCA-HWC1040
COMPARE
999 245 High Temp Heating Cord with SDC Temp Controller, 4 ft (1.2 m) Length, 245 Watts, °C Temp Display; 120 VAC
$446.00USD / Each
Showing 1- 13 of 13