Branson Sonifier® Analog 200- and 400-Watt Ultrasonic Processors
Easy-to-use dials simplify operation
  • Continuous, timed, and pulse modes provide versatility for changing applications
  • Power output control and autotuning optimize processing performance
  • Mount converter to a laboratory stand for hands-free dispersion
These analog sonicators are compact to save bench space, and feature a sloped control panel for greater visibility and easy access to controls. Power output meter allows for process monitoring and increases reproducibility among samples. Choose the ultrasonic processor with the power output and horn diameter required for your sample volume.
$4,200.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Branson 450 Analog Sonifier with 1/2" Horn, 400 W, 240 VAC EW-04715-04 Mfr # 101-063-199R
COMPARE
Analog Sonifier with 1/2\" Horn, 400 W, 240 VAC
$4,200.00USD / Each