Boekel Scientific™ Touch-Screen Refrigerated Incubators
Advanced color touch screen for custom temperature and time-based control
  • Quickly download data via USB or Ethernet connections
  • Easily add humidity to the chamber with included humidity pan
  • Constructed with thermoformed plastic interior for easy cleaning
  • Internal power outlet with time-based control options
This touch screen refrigerated incubator provides easy viewing of alarms, current temperatures, and humidity level from across the room. The 4.3” color screen displays temperature and humidity graphs which can be exported via USB and Ethernet connections. Easily control and define temperature programs, alarm setpoints, and control the internal power outlet based on time.
The incubator features thermoelectric cooling, ideal for standard microbial or low temperature work such as BOD testing, drosophila, and extremophile bacteria culture. Easily add humidity to the chamber with included humidity pan. The thermoformed plastic interior is easy to clean and can hold up to 86 BOD bottles.
$5,410.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Max Temperature (° C) Power (VAC)
Availability Pricing
Boekel Scientific 165000 Touch Screen Refrigerated Incubator, 115V EW-01100-03 Mfr # 165000
COMPARE
4 60 115
$5,410.00USD / Each