Bio-bin Waste Disposal Containers
Great alternative to plastic waste collection and disposal containers
  • Space-efficient, lightweight fold-flat design
  • Waterproof construction — inside and outside — with heat-sealed joints
  • Built-in super absorbent Bio-matt™ liner soaks up excess liquid
These disposable plastic-coated paper containers are perfect for laboratory waste that requires special heat treatment such as autoclaving or incineration before disposal. The built-in lid has both a temporary and permanent seal and features a handle for easy transport when full. Ideal for use in academic, clinical, government, and industrial laboratories for a wide range of non-sharp biohazard or non-hazardous waste. Autoclavable
$240.00 - $322.50USD / Case of 50
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters)
Availability Pricing
Dynalon 797303-0002 Bio-bin Waste Disposal Container, 2.5L, Case of 50 EW-81402-07 Mfr # 797303-0002
COMPARE
2.5
$250.00USD / Case of 50
Dynalon 797303-0006 Bio-bin Waste Disposal Container, 6L, Case of 40 EW-81402-08 Mfr # 797303-0006
COMPARE
6
$322.50USD / Case of 40
Dynalon 797303-0030 Bio-bin Waste Disposal Container, 30L, Case of 10 EW-81402-09 Mfr # 797303-0030
COMPARE
30
$240.00USD / Case of 10