Binder UFV Ultra-Low Temperature Freezers
Ensures safe, long-term storage of samples at -90°C (-130°F)
  • Lowest energy consumption in its class
  • Innovative door gasket reduces buildup of ice
  • Three stainless steel shelves provide flexible placement
  • Stainless steel finish and smooth inside surface make cleanup easy
These ultra-low temperature freezers with climate-neutral refrigerants (R-290 and R-170) offer safe, long-term storage of biological and chemical samples within a temperature range of -90 to -40°C (-130 to -40°F). The freezers guarantee a constant low temperature that can be adjusted to an accuracy of one degree. The ambient temperature is monitored by a controller, which emits a visual or acoustic alarm if a preset value is exceeded.
A multi-level safety concept protects against unauthorized access. The door can be opened using a door latch handle or an electromechanical door locking mechanism. You can also lock the temperature controller's keypad to ensure maximum security and password-protect the adjustment menus.
Additional features include an ergonomic door handle, thermal insulation with vacuum insulation panels, Ethernet interface, internal data logger, and operating hours counter.
$19,890.00 - $24,542.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Power (Hz) Power (VAC)
Availability Pricing
Binder UFV500UL-120V Standard Series Ultra-Low Temperature Freezers, 477 L; 120 VAC, 60 Hz EW-05012-78 Mfr # 9020-0351
COMPARE
16.8 60 120
$19,890.00USD / Each
Binder UFV500UL-240V Standard Series Ultra-Low Temperature Freezers, 477 L; 240 VAC, 60 Hz EW-05012-79 Mfr # 9020-0353
COMPARE
16.8 60 240
$19,890.00USD / Each
Binder UFV500-230V Standard Series Ultra-Low Temperature Freezers, 477 L; 230 VAC, 50 Hz EW-05012-81 Mfr # 9020-0347
COMPARE
16.8 50 230
$19,890.00USD / Each
Binder UFV500-230V-W Water Cooling Series Ultra-Low Temperature Freezers, 477 L; 230 VAC, 50 Hz EW-05012-82 Mfr # 9020-0349
COMPARE
16.8 50 230
$22,521.00USD / Each
Binder UFV700UL-120V Standard Series Ultra-Low Temperature Freezers, 700 L; 120 VAC, 60 Hz EW-05012-83 Mfr # 9020-0352
COMPARE
24.7 60 120
$21,909.00USD / Each
Binder UFV700UL-240V Standard Series Ultra-Low Temperature Freezers, 700 L; 240 VAC, 60 Hz EW-05012-84 Mfr # 9020-0354
COMPARE
24.7 60 240
$21,909.00USD / Each
Binder UFV700-230V Standard Series Ultra-Low Temperature Freezers, 700 L; 230 VAC, 50 Hz EW-05012-86 Mfr # 9020-0348
COMPARE
24.7 50 230
$21,909.00USD / Each
Binder UFV700-230V-W Water Cooling Series Ultra-Low Temperature Freezers, 700 L; 230 VAC, 50 Hz EW-05012-87 Mfr # 9020-0350
COMPARE
24.7 50 230
$24,542.00USD / Each
Showing 1- 8 of 8