This product is now discontinued, View Related Products

SHOWA T-Flex Cut-Resistant Gloves & Sleeves

No need to sacrifice tactile sensitivity for cut-resistant protection
Made of Dyneema®-Spectra™ engineered fibers, this lightweight glove and sleeve offer an ANSI/ ASTM Level IV cut rating. Ideal for handling box cutters, knives, shears, and fabricated and sharp edged metals that require a high level of dexterity — useful in meat and poultry processing. Choose T-Flex Plus® gloves with coated dots for increased dexterity. All are fully washable. Ambidextrous. Approval: CE
Compliance: Made with FDA compliant material for food handling
Call For Price

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Size
Availability Pricing
SHOWA 8115-07 Cut-Resistant Glove, Uncoated; size, small EW-86483-30 Mfr # 8115-07
COMPARE
Small
Call For Price
SHOWA 8115-08 Cut-Resistant Glove, Uncoated; size, Medium EW-86483-31 Mfr # 8115-08
COMPARE
Medium
Call For Price
SHOWA 8115-09 Cut-Resistant Glove, Uncoated; size, large EW-86483-32 Mfr # 8115-09
COMPARE
Large
Call For Price
SHOWA 8115-10 Cut-Resistant Glove, Uncoated; size, x-large EW-86483-33 Mfr # 8115-10
COMPARE
X-Large
Call For Price
SHOWA 8113C-07 Best T-Flex Cut-Resistant Glove, Full-Face Natural Rubber Dotted Coating; size, small EW-86483-35 Mfr # 8113C-07
COMPARE
Small
Call For Price
SHOWA 8113C-08 Best T-Flex Cut-Resistant Glove, Full-Face Natural Rubber Dotted Coating; size, medium EW-86483-36 Mfr # 8113C-08
COMPARE
Medium
Call For Price
SHOWA 8113C-09 Best T-Flex Cut-Resistant Glove, Full-Face Natural Rubber Dotted Coating; size, large EW-86483-37 Mfr # 8113C-09
COMPARE
Large
Call For Price
SHOWA 8113C-10 Best T-Flex Cut-Resistant Glove, Full-Face Natural Rubber Dotted Coating; size, x-large EW-86483-38 Mfr # 8113C-10
COMPARE
X-Large
Call For Price
SHOWA S8115M-10 Best T-Flex Cut-Resistant Sleeve, 10"; size, medium EW-86483-40 Mfr # S8115M-10
COMPARE
Medium
Call For Price
SHOWA S8115L-10 Best T-Flex Cut-Resistant Sleeve, 10"; size, large EW-86483-41 Mfr # S8115L-10
COMPARE
Large
Call For Price
SHOWA S8115M-16 Best T-Flex Cut-Resistant Sleeve, 16"; size, medium EW-86483-42 Mfr # S8115M-16
COMPARE
Medium
Call For Price
SHOWA S8115L-16 Best T-Flex Cut-Resistant Sleeve, 16"; size, large EW-86483-43 Mfr # S8115L-16
COMPARE
Large
Call For Price
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat