Bel-Art Microtube Homogenizer System Pestles and Microtubes

Process precipitates, pellets, tissue, and cells directly in the microtube!
  • Homogenize samples directly in their storage container
  • Obtain thorough mixtures with the tight pestle-to-microtube tolerance
  • Protect your samples—tubes and pestles are made from virgin polypropylene
$112.00 - $164.00USD / Pkg of 100

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Bel-Art F65000-0005 Tube and Pestle, 0.5 mL, Individually Packed; 100/Pk EW-44444-76 Mfr # F65000-0005
COMPARE
Tube and Pestle, 0.5 mL, Individually Packed; 100/Pk
$157.00USD / Pkg of 100
Bel-Art F65000-0002 Pestle, 1.5 mL, Individually Packed; 100/Pk EW-44444-78 Mfr # F65000-0002
COMPARE
Pestle, 1.5 mL, Individually Packed; 100/Pk
$143.00USD / Pkg of 100
Bel-Art F65000-0006 Tube and Pestle, 1.5 mL, Individually Packed; 100/Pk EW-44444-82 Mfr # F65000-0006
COMPARE
Tube and Pestle, 1.5 mL, Individually Packed; 100/Pk
$164.00USD / Pkg of 100
Bel-Art F65000-9002 Pestle, 1.5 mL, Bulk Packed; 100/Pk EW-44444-90 Mfr # F65000-9002
COMPARE
Pestle, 1.5 mL, Bulk Packed; 100/Pk
$112.00USD / Pkg of 100
Bel-Art F65000-9006 Tube and Pestle, 1.5 mL, Bulk Packed; 100/Pk EW-44444-94 Mfr # F65000-9006
COMPARE
Tube and Pestle, 1.5 mL, Bulk Packed; 100/Pk
$133.00USD / Pkg of 100
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat