This product is now discontinued, View Related Products
Bacharach Fyrite INSIGHT Plus Combustion Analyzer
Five-in-one instrument with full-color graphical display for all of your emission analysis needs
The Fyrite® INSIGHT® Plus is a handheld residential and commercial combustion analyzer ideal for use by HVAC contractors, home inspectors, maintenance personnel, and energy auditors. Select from nine preprogrammed fuels and measure combustion efficiency, room air carbon monoxide, flue gas carbon monoxide, two channels of temperature, and differential pressure. The B-Smart® sensor technology allows precalibrated CO sensor replacement in the field (order replacement sensor separately).
Dynamic full-color graphical display features multiple display options. The standard display shows numerical values for the parameters; the graphical trend screen displays a line graph of the selected parameter over time; the bar graph screen displays up to three parameters in bar graphs with user-defined limits; and the dynamic hot-spot screen is used to locate the hot spot in the flue, based on the stack temperature.
  • Nine preprogrammed and two custom fuels
  • Store up to 100 records for recall on the display
  • Easily navigate the menu system through the function and arrow buttons
  • Zoom function allows you to increase the character size on the display for easier viewing
  • User diagnostics include O2 sensor life indications and calibration reminders
  • Versions with reporting kit include an IrDA printer, USB cable, and software
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Includes
Availability Pricing
Bacharach Fyrite INSIGHT Plus Combustion Analyzer, 2-year sensor EW-86495-50 Mfr # 0024-8515
COMPARE
Call For Price
Bacharach Fyrite INSIGHT Plus Combustion Analyzer, long-life O2 sensor EW-86495-51 Mfr # H0024-8517
COMPARE
3-year oxygen sensor
Call For Price
Bacharach Fyrite INSIGHT Plus Combustion Analyzer with Reporting Kit EW-86495-52 Mfr # 0024-8516
COMPARE
2-year oxygen sensor and reporting kit
Call For Price
Bacharach Fyrite INSIGHT Plus Combustion Analyzer; O2, Reporting Kit EW-86495-53 Mfr # 0024-8518
COMPARE
3-year oxygen sensor and reporting kit