Bürkle Sampling Seal for Food and Pharmaceticals
Labeling and sealing conveniently combined to prevent outflow after sampling
  • Quickly seal holes from sampling lances with these sampling seals
  • Powerful adhesive ensures the seal remains firmly intact
  • Provide quality control data about the sample taken while also sealing where conventional labels will not stick
Sampling control seals for food and pharma use an adhesive approved for use with foodstuff according to FDA guidelines. These labels provide a reliable seal that will prevent the products of a container from leaking out after sampling. These labels are constructed of an aluminum back with a paper top label, creating a hermetically sealed container, ideal for use with food, pharmaceuticals, cosmetics, and even frozen goods. These labels can be written on with ball-point pen, pencil, or marker and used to provide quality control details at the time of sampling such as date, name, and contents. These labels are available in two size options and five printed color options — red, yellow, blue, green, and black — as well as a white unprinted option. Use the 06040-90 dispenser for the 95 x 95 mm labels and the 06040-91 dispenser for the larger 150 x 150 mm labels.
$111.75 - $153.75USD / Roll
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Width (in)
Availability Pricing
Burkle 5303-2017 Sampling Control Seal, FDA compliant Adhesive, 95 x 95 mm; Red EW-06040-76 Mfr # 5303-2017
COMPARE
Red 3.75 3.75
$111.75USD / Roll
Burkle 5303-2018 Sampling Control Seal, FDA Compliant Adhesive, 95 x 95 mm; White EW-06040-77 Mfr # 5303-2018
COMPARE
White 3.75 3.75
$111.75USD / Roll
Burkle 5303-2020 Sampling Control Seal, FDA Compliant Adhesive, 95 x 95 mm; Yellow EW-06040-79 Mfr # 5303-2020
COMPARE
Yellow 3.75 3.75
$111.75USD / Roll
Burkle 5303-2021 Sampling Control Seal, FDA Compliant Adhesive, 95 x 95 mm; Blue EW-06040-80 Mfr # 5303-2021
COMPARE
Blue 3.75 3.75
$111.75USD / Roll
Burkle 5303-2022 Sampling Seal Tape, FDA Compliant Adhesive, 95 x 50 m; Green EW-06040-81 Mfr # 5303-2022
COMPARE
Green 2 3.75
$111.75USD / Roll
Burkle 5303-2023 Sampling Control Seal, FDA Compliant Adhesive, 95 x 95 mm; Black EW-06040-82 Mfr # 5303-2023
COMPARE
Black 3.75 3.75
$111.75USD / Roll
Burkle 5303-3017 Sampling Control Seal, FDA Compliant Adhesive, 150 x 150 mm; Red EW-06040-83 Mfr # 5303-3017
COMPARE
Red 5.9 5.9
$153.75USD / Roll
Burkle 5303-3018 Sampling Control Seal, FDA compliant Adhesive, 150 x 150 mm; White EW-06040-84 Mfr # 5303-3018
COMPARE
White 5.9 5.9
$152.50USD / Roll
Burkle 5303-3021 Sampling Control Seal, FDA compliant Adhesive, 150 x 150 mm; Blue EW-06040-87 Mfr # 5303-3021
COMPARE
Blue 5.9 5.9
$152.50USD / Roll
Burkle 5303-3022 Sampling Control Seal, FDA Compliant Adhesive, 150 x 150 mm; Green EW-06040-88 Mfr # 5303-3022
COMPARE
Green 5.9 5.9
$152.50USD / Roll
Showing 1- 10 of 10