B&K Precision Dual Display LCR Meters
Precision component testing with the press of a button
  • 0.5% basic accuracy
  • Dual LCD display
  • Two selectable test frequencies of 120 Hz & 1 kHz (plus 100 Hz & 10 kHz model 879B)
  • Auto ranging
  • Mini USB interface
  • 3 Year warranty
These LCR meters perform quick, fully auto-ranging capacitance, resistance, and inductance measurements. Components can be measured with selectable test frequencies in series or parallel modes as desired. The 40,000 count main LCD displays the measured component value, and the secondary 10,000 count display reads D or Q (plus phase angle and ESR measurements model 879B).
$416.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Test Frequency
Availability Pricing
B&K Precision 879B Universal Deluxe LCR Meter EW-20036-83 Mfr # 879B
COMPARE
4 Selectable: 100Hz=100 Hz, 120Hz=120 Hz, 1KHz=1010 Hz, 10KHz=9.6 KHz
$416.00USD / Each