Ashcroft® Type A Stainless Steel Pressure Switches
Rugged stainless steel switches are easily field-adjustable
  • Precision snap-acting switch element
  • Factory set to switch at rising pressures
These pressure switches are designed for tough industrial and OEM applications requiring a durable, high-quality miniature switch — especially where corrosion resistance or leakage is a concern. Ideal for pressure alarm, shutdown, or control on heavy vehicles, food and beverage processing equipment, and wherever size is a consideration or equipment is being downsized. Models 68350-52 to -60 have a stainless steel actuator seal; models 68350-62 to -71 have a Buna N actuator seal. Switches set to activate at decreasing pressures are available. Contact an application specialist for more information.
$210.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Ashcroft Pressure Switch, Stainless Steel, 0 to 30 PSI; 1/4" NPT(M) EW-68350-54 Mfr # APAN41H012LS0230#-NSR
COMPARE
30
$210.00USD / Each
Ashcroft Pressure Switch, Stainless Steel, 0 to 100 PSI; 1/4" NPT(M) EW-68350-58 Mfr # APAN41H012LS02100#-NSR
COMPARE
100
$210.00USD / Each