Ashcroft® 3.5" Dry Sanitary Pressure Gauges
FDA compliant and meets USDA requirements and 3A Sanitary Standard 74-00
  • True Zero™ pointer indication — no stop pin to mask false zero reading
  • Use in sterile environments such as food, dairy, and pharmaceutical processing industries — autoclavable to 270°F (149°C)
Used typically in pharmaceutical, biotechnical, ultra-filtration, or food and dairy industries, these gauges can serve special applications involving chemicals, hard-to-handle liquids, and slurries. Gauges feature electropolished 316L stainless steel socket, Bourdon tube, and diaphragm seal; 300-series stainless steel case and ring; and polycarbonate lens. Bottom connection is 1-½" sanitary quick-connect. Glycerin-filled gauges use pharmaceutical/food quality USP grade glycerin (99.5% pure). Gauges are autoclaveable at 270°F for 15 minutes
ITEM(S) ON SALE
$366.80 - $468.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI)
Availability Pricing
Ashcroft 3.5" Dry Sanitary Pressure Gauge, 30 psig, 1.5" Tri-Clamp EW-68806-10 Mfr # 35-1032S-15L-30#
COMPARE
30
$374.80USD / Each
$468.50USD / Each
Ashcroft 3.5" Dry Sanitary Pressure Gauge, 60 psig, 1.5" Tri-Clamp EW-68806-11 Mfr # 35-1032S-15L-60#
COMPARE
60
$468.50USD / Each
Ashcroft 3.5" Dry Sanitary Pressure Gauge, 100 psig, 1.5" Tri-Clamp EW-68806-12 Mfr # 35-1032S-15L-100#
COMPARE
100
$366.80USD / Each
$458.50USD / Each