Ashcroft® Type 2084, 2086, and 2089 Precision Digital Test Gauges
Total Error Band Accuracy (includes: terminal point, hysteresis, linearity, repeatability and temperature from 0/150°F)
  • Units of measure include: psi, inHg, inH2O, ftSW, bar, mbar, kPa, MPa, mmHg, cm2O, mm2O, kg/cm2
  • Seven selectable languages: English, Spanish, French, German, Italian, Portuguese, and Dutch
The Ashcroft® 2084, 2086, and 2089 digital test gauges are highly accurate test instruments providing exceptional features and performance. The full scale total error (TEB) includes the effects of linearity, hysteresis, repeatability and temperature from 0 to 150°F. Silicone MEMS sensor technology provides accuracy and long term stability demanded by test instrumentation users. An ideal indicator for test and measurement applications.
$1,108.00 - $1,498.00USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Ashcroft 20843.0 3" Digital Test Pressure Gauge 2000 psi, 0.25% Accuracy EW-68435-40 Mfr # 30-2084-SD-02L-2000#
COMPARE
3\" Digital Test Pressure Gauge 2000 psi, 0.25% Accuracy
$1,108.00USD / Each
Ashcroft 20843.0 3" Digital Test Pressure Gauge 30 psi, 0.25% Accuracy EW-68435-41 Mfr # 30-2084-SD-02L-30#
COMPARE
3\" Digital Test Pressure Gauge 30 psi, 0.25% Accuracy
$1,108.00USD / Each
Ashcroft 20843.0 3" Digital Test Pressure Gauge 300 psi, 0.25% Accuracy EW-68435-42 Mfr # 30-2084-SD-02L-300#
COMPARE
3\" Digital Test Pressure Gauge 300 psi, 0.25% Accuracy
$1,108.00USD / Each
Ashcroft 20843.0 3" Digital Test Pressure Gauge 5 psi, 0.25% Accuracy EW-68435-44 Mfr # 30-2084-SD-02L-5#
COMPARE
3\" Digital Test Pressure Gauge 5 psi, 0.25% Accuracy
$1,108.00USD / Each
Ashcroft 20843.0 3" Digital Test Pressure Gauge 5000 psi, 0.25% Accuracy EW-68435-45 Mfr # 30-2084-SD-02L-5000#
COMPARE
3\" Digital Test Pressure Gauge 5000 psi, 0.25% Accuracy
$1,108.00USD / Each
Ashcroft 20863.0 3" Digital Test Pressure Gauge 15 psi, 0.1% Accuracy EW-68435-52 Mfr # 30-2086-SD-02L-15#
COMPARE
3\" Digital Test Pressure Gauge 15 psi, 0.1% Accuracy
$1,292.00USD / Each
Ashcroft 20863.0 3" Digital Test Pressure Gauge 30 psi, 0.1% Accuracy EW-68435-58 Mfr # 30-2086-SD-02L-30#
COMPARE
3\" Digital Test Pressure Gauge 30 psi, 0.1% Accuracy
$1,292.00USD / Each
Ashcroft 20863.0 3" Digital Test Pressure Gauge 5 psi, 0.1% Accuracy EW-68435-61 Mfr # 30-2086-SD-02L-5#
COMPARE
3\" Digital Test Pressure Gauge 5 psi, 0.1% Accuracy
$1,292.00USD / Each
Ashcroft 20863.0 3" Digital Test Pressure Gauge 10 psi, 0.05% Accuracy EW-68435-66 Mfr # 30-2089-SD-02L-10#
COMPARE
3\" Digital Test Pressure Gauge 10 psi, 0.05% Accuracy
$1,498.00USD / Each
Ashcroft 20893.0 3" Digital Test Pressure Gauge 160 psi, 0.05% Accuracy EW-68435-69 Mfr # 30-2089-SD-02L-160#
COMPARE
3\" Digital Test Pressure Gauge 160 psi, 0.05% Accuracy
$1,498.00USD / Each
Ashcroft 20893.0 3" Digital Test Pressure Gauge 200 psi, 0.05% Accuracy EW-68435-70 Mfr # 30-2089-SD-02L-200#
COMPARE
3\" Digital Test Pressure Gauge 200 psi, 0.05% Accuracy
$1,498.00USD / Each
Ashcroft 20893.0 3" Digital Test Pressure Gauge 300 psi, 0.05% Accuracy EW-68435-72 Mfr # 30-2089-SD-02L-300#
COMPARE
3\" Digital Test Pressure Gauge 300 psi, 0.05% Accuracy
$1,498.00USD / Each
Ashcroft 20893.0 3" Digital Test Pressure Gauge 5 psi, 0.05% Accuracy EW-68435-73 Mfr # 30-2089-SD-02L-5#
COMPARE
3\" Digital Test Pressure Gauge 5 psi, 0.05% Accuracy
$1,498.00USD / Each
Showing 1- 13 of 13