Ashcroft® K1 Pressure Transducers
Long-term stability and repeatability
  • 0.5% and 1.0% terminal point based accuracy
  • Vac-20,000 psi pressure range
  • FM and UL options available
  • Superior long term stability and repeatability
  • Stainless steel NEMA 4X enclosure
  • Current / voltage output
  • Wide range of electrical and process connections available
Applications include hydraulic, refrigeration, machine tool, test/measurement, pump control, construction equipment, general industrial and OEM. This transmitter offers the benefits of polysilicon thin film performance at affordable prices. Using chemical vapor deposition methods polysilicon strain gauges are molecularly bonded to a metal diaphragm. This sensor, combined with the latest surface mount electronics promote superior performance in a durable package.
$398.00 - $501.50USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Ashcroft K1 Pressure Transducer, 1/4" NPT(M) Connection, 0 to 100 psi, 0.5% Accuracy, 4-20 mA EW-68440-26 Mfr # K1-5-M02-42-F2-100#
COMPARE
Pressure Transducer, 1/4" NPT(M) Connection, 0 to 100 psi, 0.5% Accuracy, 4-20 mA
$501.50USD / Each
Ashcroft K1 Pressure Transducer, 1/4" NPT(M) Connection, 0 to 150 psi, 0.5% Accuracy, 4-20 mA EW-68440-27 Mfr # K1-5-M02-42-F2-150#
COMPARE
Pressure Transducer, 1/4" NPT(M) Connection, 0 to 150 psi, 0.5% Accuracy, 4-20 mA
$501.50USD / Each
Ashcroft K1 Pressure Transducer, 1/4" NPT(M) Connection, 0 to 2000 psi, 0.5% Accuracy, 4-20 mA EW-68440-28 Mfr # K1-5-M02-42-F2-2000#
COMPARE
Pressure Transducer, 1/4" NPT(M) Connection, 0 to 2000 psi, 0.5% Accuracy, 4-20 mA
$501.50USD / Each
Ashcroft K1 Pressure Transducer, 1/4" NPT(M) Connection, 0 to 200 psi, 0.5% Accuracy, 4-20 mA EW-68440-29 Mfr # K1-5-M02-42-F2-200#
COMPARE
Pressure Transducer, 1/4" NPT(M) Connection, 0 to 200 psi, 0.5% Accuracy, 4-20 mA
$501.50USD / Each
Ashcroft K1 Pressure Transducer, 1/4" NPT(M) Connection, 0 to 300 psi, 0.5% Accuracy, 4-20 mA EW-68440-31 Mfr # K1-5-M02-42-F2-300#
COMPARE
Pressure Transducer, 1/4" NPT(M) Connection, 0 to 300 psi, 0.5% Accuracy, 4-20 mA
$501.50USD / Each
Ashcroft K1 Pressure Transducer, 1/4" NPT(M) Connection, 0 to 5000 psi, 0.5% Accuracy, 4-20 mA EW-68440-32 Mfr # K1-5-M02-42-F2-5000#
COMPARE
Pressure Transducer, 1/4" NPT(M) Connection, 0 to 5000 psi, 0.5% Accuracy, 4-20 mA
$501.50USD / Each
Ashcroft K1 Pressure Transducer, 1/4" NPT(M) Connection, 30 to 0" Hg, 1.0% Accuracy, 4-20 mA EW-68440-34 Mfr # K1-7-M02-42-F2-VAC/0#
COMPARE
Pressure Transducer, 1/4" NPT(M) Connection, 30 to 0" Hg, 1.0% Accuracy, 4-20 mA
$415.00USD / Each
Ashcroft K1 Pressure Transducer, 1/4" NPT(M) Connection, 0 to 100 psi, 1.0% Accuracy, 4-20 mA EW-68440-36 Mfr # K1-7-M02-42-F2-100#
COMPARE
Pressure Transducer, 1/4" NPT(M) Connection, 0 to 100 psi, 1.0% Accuracy, 4-20 mA
$398.00USD / Each
Ashcroft K1 Pressure Transducer, 1/4" NPT(M) Connection, 0 to 150 psi, 1.0% Accuracy, 4-20 mA EW-68440-37 Mfr # K1-7-M02-42-F2-150#
COMPARE
Pressure Transducer, 1/4" NPT(M) Connection, 0 to 150 psi, 1.0% Accuracy, 4-20 mA
$398.00USD / Each
Ashcroft K1 Pressure Transducer, 1/4" NPT(M) Connection, 0 to 15 psi, 1.0% Accuracy, 4-20 mA EW-68440-38 Mfr # K1-7-M02-42-F2-15#
COMPARE
Pressure Transducer, 1/4" NPT(M) Connection, 0 to 15 psi, 1.0% Accuracy, 4-20 mA
$415.00USD / Each
Ashcroft K1 Pressure Transducer, 1/4" NPT(M) Connection, 0 to 2000 psi, 1.0% Accuracy, 4-20 mA EW-68440-40 Mfr # K1-7-M02-42-F2-2000#
COMPARE
Pressure Transducer, 1/4" NPT(M) Connection, 0 to 2000 psi, 1.0% Accuracy, 4-20 mA
$398.00USD / Each
Ashcroft K1 Pressure Transducer, 1/4" NPT(M) Connection, 0 to 200 psi, 1.0% Accuracy, 4-20 mA EW-68440-41 Mfr # K1-7-M02-42-F2-200#
COMPARE
Pressure Transducer, 1/4" NPT(M) Connection, 0 to 200 psi, 1.0% Accuracy, 4-20 mA
$398.00USD / Each
Ashcroft K1 Pressure Transducer, 7/16"-20 SAE(M) Connection, 0 to 3000 psi, 1.0% Accuracy, 1-6 VDC EW-68440-50 Mfr # K1-7-MEK-16-HM-3000#
COMPARE
Pressure Transducer, 7/16"-20 SAE(M) Connection, 0 to 3000 psi, 1.0% Accuracy, 1-6 VDC
$412.00USD / Each
Showing 1- 13 of 13