Ashcroft® DG25 Digital Pressure Gauges with Backlight
Stainless steel sensor and nine engineering units make this gauge suitable for many applications
  • ±0.5% full-scale accuracy
  • IP67 rated case
  • Easy-to-read, 5-digit display with 20-segment bar graph
A versatile and economic choice for a wide variety of pressure measurement applications. Gauges feature easy-to-use, three-button keypad programming. Cycle life of 108 makes these gauges a great value.
Case material is made of PC/ABS. Gauges feature tare, min/max memory, and nine engineering plus one custom engineering units, as well as a bright backlight. Sensor is welded to the socket for added durability. Max pressure indication reduces chances of damage to the unit. Burst pressure rating is 300 to 800% of range. Gauges meet ASME B40.7 safety requirement. Other pressure connections, such as 1/8" NPT(M), are available. Call for assistance.
ITEM(S) ON SALE
$300.00 - $375.00USD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Accuracy Resolution Description
Availability Pricing
Ashcroft DG25 Digital Pressure Gauge with Backlight, Vacuum; 0 to 30" Hg
Always in Stock
EW-68334-01 Mfr # DG2551L0NAM02L0IM&V-XZZ
COMPARE
±0.5% full-scale 0.01\" Hg Digital Vacuum Gauge with Backlight, 0-30\"Hg
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, Compound; 30" Hg, 30 psi EW-68334-03 Mfr # DG2551L0NAM02L30#&V-XZZ
COMPARE
±0.5% full-scale 0.01\" Hg Digital Compound Gauge with Backlight, 30\"Hg-30 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, Compound; 30" Hg, 60 psi EW-68334-05 Mfr # DG2551L0NAM02L60#&V-XZZ
COMPARE
±0.5% full-scale 0.01\" Hg Digital Compound Gauge with Backlight, 30\"Hg-60 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, Compound; 30" Hg, 100 psi
Always in Stock
EW-68334-07 Mfr # DG2551L0NAM02L100#&V-XZZ
COMPARE
±0.5% full-scale 0.1\" Hg Digital Compound Gauge with Backlight, 30\"Hg-100 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 15 psi EW-68334-09 Mfr # DG2551L0NAM02L15#-XZZ
COMPARE
±0.5% full-scale 0.001 psi Digital Pressure Gauge with Backlight, 0-15 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 30 psi EW-68334-11 Mfr # DG2551L0NAM02L30#-XZZ
COMPARE
±0.5% full-scale 0.01 psi Digital Pressure Gauge with Backlight, 0-30 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 60 psi EW-68334-13 Mfr # DG2551L0NAM02L60#-XZZ
COMPARE
±0.5% full-scale 0.01 psi Digital Pressure Gauge with Backlight, 0-60 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 100 psi EW-68334-15 Mfr # DG2551L0NAM02L100#-XZZ
COMPARE
±0.5% full-scale 0.01 psi Digital Pressure Gauge with Backlight, 0-100 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 200 psi
Always in Stock
EW-68334-17 Mfr # DG2551L0NAM02L200#-XZZ
COMPARE
±0.5% full-scale 0.01 psi Digital Pressure Gauge with Backlight, 0-200 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 300 psi
Always in Stock
EW-68334-19 Mfr # DG2551L0NAM02L300#-XZZ
COMPARE
±0.5% full-scale 0.1 psi Digital Pressure Gauge with Backlight, 0-300 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 500 psi
Always in Stock
EW-68334-21 Mfr # DG2551L0NAM02L500#-XZZ
COMPARE
±0.5% full-scale 1 psi Digital Pressure Gauge with Backlight, 0-500 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 750 psi EW-68334-23 Mfr # DG2551L0NAM02L750#-XZZ
COMPARE
±0.5% full-scale 0.1 psi Digital Pressure Gauge with Backlight, 0-750 psi
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 1000 psi EW-68334-25 Mfr # DG2551L0NAM02L1000#-XZZ
COMPARE
±0.5% full-scale 0.1 psi Digital Pressure Gauge with Backlight, 0-1000 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 5000 psi EW-68334-27 Mfr # DG2551L0NAM02L5000#-XZZ
COMPARE
±0.5% full-scale 1 psi Digital Pressure Gauge with Backlight, 0-5000 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 7500 psi EW-68334-29 Mfr # DG2551L0NAM02L7500#-XZZ
COMPARE
±0.5% full-scale 1 psi Digital Pressure Gauge with Backlight, 0-7500 psi
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 10000 psi
Always in Stock
EW-68334-31 Mfr # DG2551L0NAM02L10000#-XZZ
COMPARE
±0.5% full-scale 1 psi Digital Pressure Gauge with Backlight, 0-10000 psi
$314.80USD / Each
$393.50USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 20000 psi EW-68334-33 Mfr # DG2551L0NAM02L20000#-XZZ
COMPARE
±0.5% full-scale 1 psi Digital Pressure Gauge with Backlight, 0-20000 psi
$300.00USD / Each
$375.00USD / Each
Ashcroft DG25 Digital Pressure Gauge with Backlight, Compound; 30" Hg to 300 psi EW-68334-35 Mfr # DG2551L0NAM02L300#&V-XZZ
COMPARE
±0.5% full-scale 0.1\" Hg Digital Compound Gauge with Backlight, 30\"Hg-300 psi
$300.00USD / Each
$375.00USD / Each
Showing 1- 18 of 18