Ashcroft® Commercial Gauges with Back Connection
Gauges feature PowerflexX™ shock and vibration protection
  • True zero indication identifies damaged gauges — increases safety and ensures accuracy
Use these glycerin-filled gauges for easier reading when high-frequency pressure fluctuations or vibrations are present. Gauges construction includes brass socket, phosphor bronze tube, 304 stainless steel case, and plastic lens.
ITEM(S) ON SALE
$29.60 - $33.60USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Accuracy Description
Availability Pricing
Ashcroft 30052.5 Commercial Gauge with Back Connection, 0 to 15 psi, 2-1/2" Dial EW-68008-02 Mfr # 63-W-1008-A-L-02B-15#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Back Connection, 0 to 15 psi, 2-1/2\" Dial
$29.60USD / Each
$37.00USD / Each
Ashcroft 30052.5 Commercial Gauge with Back Connection, 0 to 30 psi, 2-1/2" Dial EW-68008-03 Mfr # 63-1008-A-L-02B-30#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Back Connection, 0 to 30 psi, 2-1/2\" Dial
$31.20USD / Each
$39.00USD / Each
Ashcroft 30052.5 Commercial Gauge with Back Connection, 0 to 60 psi, 2-1/2" Dial EW-68008-04 Mfr # 63-W-1008-A-L-02B-60#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Back Connection, 0 to 60 psi, 2-1/2\" Dial
$31.20USD / Each
$39.00USD / Each
Ashcroft 30052.5 Commercial Gauge with Back Connection, 0 to 100 psi, 2-1/2" Dial EW-68008-05 Mfr # 63-1008-A-L-02B-100#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Back Connection, 0 to 100 psi, 2-1/2\" Dial
$31.20USD / Each
$39.00USD / Each
Ashcroft 30052.5 Commercial Gauge with Back Connection, 0 to 160 psi, 2-1/2" Dial EW-68008-06 Mfr # 63-W-1008-A-L-02B-160#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Back Connection, 0 to 160 psi, 2-1/2\" Dial
$31.20USD / Each
$39.00USD / Each
Ashcroft 30052.5 Commercial Gauge with Back Connection, 0 to 200 psi, 2-1/2" Dial EW-68008-07 Mfr # 63-W-1008-A-L-02B-200#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Back Connection, 0 to 200 psi, 2-1/2\" Dial
$31.20USD / Each
$39.00USD / Each
Ashcroft 30052.5 Commercial Gauge with Back Connection, 0 to 1000 psi, 2-1/2" Dial EW-68008-09 Mfr # 63-W-1008-A-L-02B-1000#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Back Connection, 0 to 1000 psi, 2-1/2\" Dial
$33.60USD / Each
$42.00USD / Each
Ashcroft 30052.5 Commercial Gauge with Back Connection, 0 to 3000 psi, 2-1/2" Dial EW-68008-11 Mfr # 63-W-1008-A-L-02B-3000#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Back Connection, 0 to 3000 psi, 2-1/2\" Dial
$33.60USD / Each
$42.00USD / Each
Showing 1- 8 of 8